siRNA Details
| Gene Symbol: FAM189B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267608.1 | NP_001254537.1 | 7 | 1458 | ||
| NM_006589.2 | NP_006580.2 | 8 | 1512 | |||
| NM_198264.1 | NP_937995.1 | 5 | 1224 | |||
| XM_005244845.1 | XP_005244902.1 | 8 | 1509 | |||
| GenBank mRNA | AL353941.1 | - | 3 | 188 | ||
| BC006493.2 | - | 8 | 992 | 2966515 | ||
| BC007444.2 | - | 8 | 992 | 2966515 | ||
| BC010471.1 | - | 8 | 1416 | 4179576 | ||
| BC040136.1 | - | 5 | 701 | 6061151 | ||

FAM189B
Genomic Map