siRNA Details
Gene Symbol: KLRA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028045.1 | 4 | 611 | |||
GenBank mRNA | AF047445.1 | - | 4 | 341 | ||
AK290077.1 | - | 3 | 312 | |||
AK296103.1 | - | 4 | 348 | |||
AK304677.1 | - | 3 | 307 | |||
AY334570.1 | - | 2 | 155 | |||
BC069672.1 | - | 4 | 314 | 7262588 | ||
BC069697.1 | - | 4 | 314 | 7262600 | ||
BC069734.1 | - | 4 | 314 | 7262612 | ||
BC101471.1 | - | 4 | 328 | 8068977 | ||
BC101473.1 | - | 4 | 331 | 8068979 |
