siRNA Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286132.1 | NP_001273061.1 | 1 | 105 | ||
| NM_001286133.1 | NP_001273062.1 | 1 | 105 | |||
| NM_006638.3 | NP_006629.2 | 1 | 105 | |||
| XM_011514253.2 | XP_011512555.1 | 1 | 100 | |||
| GenBank mRNA | BC017871.1 | - | 1 | 79 | 4278825 | |
| BG430780.1 | - | 1 | 80 | |||
| BM015573.1 | - | 68 | ||||
| BM827175.1 | - | 1 | 53 | |||
| BQ220155.1 | - | 1 | 53 | |||
| BX951376.1 | - | 1 | 105 | |||

RPP40
Genomic Map