siRNA Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021116.2 | NP_066939.1 | 10 | 1884 | ||
| XM_005249584.3 | XP_005249641.1 | 10 | 1882 | |||
| GenBank mRNA | AB209496.1 | - | 10 | 1829 | ||
| L05500.1 | - | 8 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY1| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021116.2 | NP_066939.1 | 10 | 1884 | ||
| XM_005249584.3 | XP_005249641.1 | 10 | 1882 | |||
| GenBank mRNA | AB209496.1 | - | 10 | 1829 | ||
| L05500.1 | - | 8 | 1028 | |||