siRNA Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021116.2 | NP_066939.1 | 20 | 3166 | ||
| GenBank mRNA | AB209496.1 | - | 20 | 3112 | ||
| BU181704.1 | - | 1 | 99 | |||
| L05500.1 | - | 18 | 2310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY1| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021116.2 | NP_066939.1 | 20 | 3166 | ||
| GenBank mRNA | AB209496.1 | - | 20 | 3112 | ||
| BU181704.1 | - | 1 | 99 | |||
| L05500.1 | - | 18 | 2310 | |||