siRNA Details
| Gene Symbol: NEU3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006656.5 | NP_006647.3 | 3 | 1448 | ||
| GenBank mRNA | AB008185.2 | - | 4 | 1363 | ||
| AK022450.1 | - | 3 | 1442 | |||
| AK290442.1 | - | 3 | 1410 | |||
| AK303540.1 | - | 2 | 1196 | |||
| BC136397.1 | - | 4 | 1782 | 9020384 | ||
| BC144059.1 | - | 4 | 1782 | 9052579 | ||
| Y18563.1 | - | 3 | 2219 | |||

NEU3
Genomic Map