siRNA Details
| Gene Symbol: EDDM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006683.4 | NP_006674.2 | 2 | 402 | ||
| XM_017020934.1 | XP_016876423.1 | 2 | 419 | |||
| GenBank mRNA | AA769825.1 | - | 1 | 476 | ||
| BC069407.1 | - | 2 | 373 | 7262138 | ||
| BC096698.1 | - | 2 | 392 | 40014618 | ||
| BC096731.1 | - | 2 | 392 | 40014622 | ||
| BC098164.1 | - | 1 | 392 | 40014621 | ||
| BC098356.1 | - | 1 | 392 | 40014616 | ||
| BC106711.1 | - | 1 | 356 | 40009240 | ||
| BC106712.1 | - | 1 | 356 | 40009242 | ||
| X76383.2 | - | 2 | 402 | |||

EDDM3A
Genomic Map