siRNA Details
| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 6, 7 | 729 | ||
| GenBank mRNA | AF083217.1 | - | 6, 7 | 710 | ||
| AK292438.1 | - | 6, 7 | 724 | |||
| BC058836.1 | - | 7, 8 | 768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR3| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 6, 7 | 729 | ||
| GenBank mRNA | AF083217.1 | - | 6, 7 | 710 | ||
| AK292438.1 | - | 6, 7 | 724 | |||
| BC058836.1 | - | 7, 8 | 768 | |||