siRNA Details
| Gene Symbol: TRAFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143906.1 | NP_001137378.1 | 8 | 1654 | ||
| NM_006700.2 | NP_006691.1 | 8 | 1123 | |||
| GenBank mRNA | AB007447.1 | - | 8 | 1091 | ||
| AK122620.1 | - | 8 | 1638 | |||
| AK291331.1 | - | 8 | 1108 | |||
| AK295474.1 | - | 8 | 864 | |||
| AK312601.1 | - | 8 | 1123 | |||
| BC003553.2 | - | 7 | 1049 | 3547039 | ||

TRAFD1
Genomic Map