siRNA Details
| Gene Symbol: TRAFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143906.1 | NP_001137378.1 | 5 | 961 | ||
| NM_006700.2 | NP_006691.1 | 5 | 430 | |||
| GenBank mRNA | AB007447.1 | - | 5 | 398 | ||
| AK122620.1 | - | 5 | 946 | |||
| AK291331.1 | - | 5 | 415 | |||
| AK295474.1 | - | 5 | 415 | |||
| AK309916.1 | - | 5 | 437 | |||
| AK312601.1 | - | 5 | 430 | |||
| BC003553.2 | - | 4 | 356 | 3547039 | ||

TRAFD1
Genomic Map