siRNA Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130914.1 | NP_001124386.1 | 5 | 729 | ||
| NM_006806.4 | NP_006797.3 | 4 | 597 | |||
| XM_011529441.1 | XP_011527743.1 | 6 | 837 | |||
| XM_017028265.1 | XP_016883754.1 | 5 | 735 | |||
| XM_017028266.1 | XP_016883755.1 | 5 | 692 | |||
| GenBank mRNA | AK289408.1 | - | 4 | 545 | ||
| BC011957.1 | - | 5 | 623 | 3859781 | ||
| BT007276.1 | - | 4 | 469 | |||
| CR457061.1 | - | 3 | 337 | |||
| D64110.1 | - | 4 | 431 | |||
| DR155504.1 | - | 4 | 513 | |||

BTG3
Genomic Map