siRNA Details
| Gene Symbol: CKAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006825.3 | NP_006816.2 | 2 | 1583 | ||
| GenBank mRNA | AK123693.1 | - | 3 | 1413 | ||
| AK223563.1 | - | 2 | 1637 | |||
| BC025341.1 | - | 3 | 1374 | |||
| BC067357.1 | - | 3 | 1493 | |||
| BC082972.1 | - | 2 | 1556 | 6084968 | ||
| BC094824.1 | - | 3 | 1364 | 6374931 | ||
| X69910.2 | - | 2 | 1583 | |||

CKAP4
Genomic Map