siRNA Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012216.3 | NP_036348.2 | 10 | 2648 | ||
| NM_052817.2 | NP_438112.2 | 10 | 2558 | |||
| XM_005262062.4 | XP_005262119.1 | 10 | 6911 | |||
| XM_017029239.1 | XP_016884728.1 | 10 | 6821 | |||
| GenBank mRNA | AF196481.1 | - | 10 | 1998 | ||
| AY625004.1 | - | 9 | 1925 | |||
| BC017707.1 | - | 9 | 1952 | 4446324 | ||
| BT006663.1 | - | 9 | 1925 | |||
| Y18880.1 | - | 10 | 2272 | |||

MID2
Genomic Map