siRNA Details
| Gene Symbol: AKR1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001818.3 | NP_001809.3 | 3, 4 | 409 | ||
| GenBank mRNA | AB031085.1 | - | 3, 4 | 376 | ||
| AB045829.1 | - | 3, 4 | 376 | |||
| AK314988.1 | - | 3, 4 | 450 | |||
| BC020744.1 | - | 3, 4 | 409 | 4734943 | ||
| D26125.1 | - | 3, 4 | 372 | |||
| M33375.1 | - | 3, 4 | 374 | |||
| S68287.1 | - | 3, 4 | 383 | |||

AKR1C4
Genomic Map