siRNA Details
| Gene Symbol: BTN2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197233.2 | NP_001184162.1 | 3 | 729 | ||
| NM_001197234.2 | NP_001184163.1 | 4 | 841 | |||
| NM_007049.4 | NP_008980.1 | 4 | 841 | |||
| NM_078476.3 | NP_510961.1 | 4 | 841 | |||
| GenBank mRNA | AB209777.1 | - | 4 | 777 | ||
| AK297098.1 | - | 3 | 683 | |||
| BC016661.1 | - | 4 | 784 | 4340392 | ||
| BX538050.1 | - | 3 | 384 | |||
| U90142.1 | - | 4 | 804 | |||
| U90543.1 | - | 4 | 782 | |||

BTN2A1
Genomic Map