siRNA Details
| Gene Symbol: KIF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300791.1 | NP_001287720.1 | 5, 6 | 781 | ||
| NM_001300792.1 | NP_001287721.1 | 5, 6 | 781 | |||
| NM_007054.6 | NP_008985.3 | 5, 6 | 781 | |||
| XM_006714526.3 | XP_006714589.1 | 5, 6 | 836 | |||
| XM_017008995.1 | XP_016864484.1 | 5, 6 | 836 | |||
| XM_017008996.1 | XP_016864485.1 | 5, 6 | 836 | |||
| GenBank mRNA | AB209780.1 | - | 5, 6 | 729 | ||
| AF041853.1 | - | 4, 5 | 622 | |||
| AK295089.1 | - | 5, 6 | 739 | |||
| AK313359.1 | - | 5, 6 | 749 | |||
| AM177178.1 | - | 4, 5 | 622 | |||
| BC020890.1 | - | 5, 6 | 746 | |||
| BC032599.1 | - | 5, 6 | 764 | |||
| BC039592.1 | - | 5, 6 | 738 | |||
| BC045542.1 | - | 5, 6 | 775 | 5298675 | ||

KIF3A
Genomic Map