siRNA Details
| Gene Symbol: C14orf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007176.3 | NP_009107.1 | 4, 5 | 803 | ||
| GenBank mRNA | AF134159.2 | - | 4, 5 | 423 | ||
| AF136971.1 | - | 4, 5 | 463 | |||
| AF161406.1 | - | 4, 5 | 463 | |||
| AK311764.1 | - | 4, 5 | 443 | |||
| AL136658.1 | - | 4, 5 | 461 | |||
| BC002444.2 | - | 4, 5 | 470 | 3347242 | ||
| BX248022.1 | - | 4, 5 | 797 | |||
| CR533443.1 | - | 3, 4 | 351 | |||

C14orf1
Genomic Map