siRNA Details
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007217.3 | NP_009148.2 | 6, 7 | 795 | ||
NM_145859.1 | NP_665858.1 | 6, 7 | 644 | |||
NM_145860.1 | NP_665859.1 | 5, 6 | 550 | |||
XM_005247086.4 | XP_005247143.1 | 6, 7 | 1530 | |||
XM_005247087.4 | XP_005247144.1 | 6, 7 | 1535 | |||
XM_005247088.3 | XP_005247145.1 | 5, 6 | 626 | |||
XM_006713485.3 | XP_006713548.1 | 6, 7 | 690 | |||
XM_011512368.2 | XP_011510670.1 | 7, 8 | 1603 | |||
XM_011512369.2 | XP_011510671.1 | 7, 8 | 1608 | |||
XM_017005644.1 | XP_016861133.1 | 6, 7 | 685 | |||
XM_017005645.1 | XP_016861134.1 | 6, 7 | 1526 | |||
GenBank mRNA | AF022385.1 | - | 5, 6 | 550 | ||
AK291130.1 | - | 5, 6 | 581 | |||
BC002506.1 | - | 6, 7 | 807 | 3050540 | ||
BC016353.1 | - | 6, 7 | 648 | 4092244 | ||
CR457107.1 | - | 4, 5 | 397 |
