siRNA Details
| Gene Symbol: RNF139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007218.3 | NP_009149.2 | 2 | 1101 | ||
| XM_017013002.1 | XP_016868491.1 | 3 | 791 | |||
| GenBank mRNA | AF064801.1 | - | 2 | 995 | ||
| AK001602.1 | - | 2 | 804 | |||
| AK025043.1 | - | 1 | 310 | |||
| AK098519.1 | - | 1 | 175 | |||
| BC021571.1 | - | 2 | 972 | 4550103 | ||
| BC064636.1 | - | 2 | 968 | 5748185 | ||

RNF139
Genomic Map