siRNA Details
| Gene Symbol: SAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320847.1 | NP_001307776.1 | 1, 2 | 295 | ||
| NM_133491.4 | NP_597998.1 | 1, 2 | 295 | |||
| GenBank mRNA | AF348524.1 | - | 1, 2 | 54 | ||
| BC011751.2 | - | 1, 2 | 265 | 3534187 | ||
| BG748008.1 | - | 1, 2 | 222 | |||

SAT2
Genomic Map