siRNA Details
| Gene Symbol: CHN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039936.2 | NP_001035025.1 | 5 | 1090 | ||
| NM_001293069.1 | NP_001279998.1 | 12 | 1361 | |||
| NM_001293070.1 | NP_001279999.1 | 12 | 1575 | |||
| NM_001293071.1 | NP_001280000.1 | 10 | 1480 | |||
| NM_001293072.1 | NP_001280001.1 | 11 | 1293 | |||
| NM_001293073.1 | NP_001280002.1 | 5 | 955 | |||
| NM_001293076.1 | NP_001280005.1 | 4 | 916 | |||
| NM_001293081.1 | NP_001280010.1 | 5 | 605 | |||
| NM_004067.3 | NP_004058.1 | 11 | 1536 | |||
| NR_120525.1 | 3 | 696 | ||||
| XM_011515105.2 | XP_011513407.1 | 14 | 1416 | |||
| XM_011515106.2 | XP_011513408.1 | 13 | 1377 | |||
| XM_011515107.2 | XP_011513409.1 | 13 | 2159 | |||
| XM_017011721.1 | XP_016867210.1 | 15 | 1434 | |||
| XM_017011722.1 | XP_016867211.1 | 14 | 2177 | |||
| GenBank mRNA | AK026415.1 | - | 5 | 1090 | ||
| AK293959.1 | - | 12 | 1155 | |||
| AK294032.1 | - | 11 | 1293 | |||
| AK294216.1 | - | 5 | 605 | |||
| AK294271.1 | - | 12 | 1362 | |||
| AK301235.1 | - | 5 | 614 | |||
| AK313021.1 | - | 11 | 1310 | |||
| AK316030.1 | - | 5 | 614 | |||
| BC112155.1 | - | 11 | 1281 | 8327623 | ||
| EU732751.1 | - | 3 | 297 | |||
| EU732752.1 | - | 5 | 556 | |||
| EU732754.1 | - | 4 | 517 | |||
| EU732760.1 | - | 10 | 1043 | |||
| EU732762.1 | - | 8 | 667 | |||
| GQ924106.1 | - | 12 | 1362 | |||
| U28926.1 | - | 11 | 1333 | |||

CHN2
Genomic Map