siRNA Details
| Gene Symbol: MGAT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014275.4 | NP_055090.1 | 10 | 1503 | ||
| NM_054013.3 | NP_463459.1 | 9 | 1943 | |||
| XM_017008998.1 | XP_016864487.1 | 10 | 1329 | |||
| GenBank mRNA | AB000624.1 | - | 10 | 1176 | ||
| AK023137.1 | - | 9 | 1943 | |||
| AL162067.1 | - | 7 | 607 | |||
| AY358984.1 | - | 10 | 1184 | |||
| BC009464.1 | - | 8 | 830 | |||
| BC051835.1 | - | 10 | 1379 | 6196058 | ||

MGAT4B
Genomic Map