siRNA Details
| Gene Symbol: CDCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080668.3 | NP_542399.1 | 5 | 716 | ||
| XM_005273733.3 | XP_005273790.1 | 5 | 711 | |||
| XM_005273734.3 | XP_005273791.1 | 5 | 711 | |||
| XM_011544743.2 | XP_011543045.1 | 5 | 711 | |||
| XM_011544748.2 | XP_011543050.1 | 5 | 711 | |||
| XM_011544749.2 | XP_011543051.1 | 5 | 711 | |||
| XM_017017158.1 | XP_016872647.1 | 5 | 711 | |||
| GenBank mRNA | AK093670.1 | - | 1 | 291 | ||
| AK291490.1 | - | 5 | 640 | |||
| AL832526.1 | - | 4 | 433 | |||
| BC011000.2 | - | 5 | 624 | 3938081 | ||

CDCA5
Genomic Map