siRNA Details
| Gene Symbol: ADAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138422.2 | NP_612431.2 | 2 | 724 | ||
| GenBank mRNA | BC011824.1 | - | 2 | 678 | 3830276 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADAT3| Gene Symbol: ADAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138422.2 | NP_612431.2 | 2 | 724 | ||
| GenBank mRNA | BC011824.1 | - | 2 | 678 | 3830276 | |