siRNA Details
| Gene Symbol: MGAT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012214.2 | NP_036346.1 | 2 | 336 | ||
| GenBank mRNA | AB000616.1 | - | 1 | 157 | ||
| AK291533.1 | - | 1 | 146 | |||
| AK314024.1 | - | 2 | 300 | |||
| BC127107.1 | - | 1 | 234 | 40124731 | ||
| BC127108.1 | - | 1 | 234 | 40124741 | ||

MGAT4A
Genomic Map