siRNA Details
| Gene Symbol: CTRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007272.2 | NP_009203.2 | 4, 5 | 386 | ||
| XM_011540550.1 | XP_011538852.1 | 4, 5 | 386 | |||
| GenBank mRNA | AK289447.1 | - | 4, 5 | 385 | ||
| BC015118.1 | - | 4, 5 | 392 | 3950170 | ||
| BC059779.1 | - | 4, 5 | 365 | |||
| CR749296.1 | - | 2, 3 | 196 | |||
| S82198.1 | - | 4, 5 | 363 | |||
| Y13697.1 | - | 3, 4 | 315 | |||

CTRC
Genomic Map