siRNA Details
| Gene Symbol: PDAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014891.6 | NP_055706.1 | 4 | 453 | ||
| GenBank mRNA | BC000684.2 | - | 4 | 365 | 3347335 | |
| BC007873.2 | - | 4 | 329 | 4303959 | ||
| BC071993.1 | - | 4 | 363 | |||
| BG281467.1 | - | 4 | 451 | |||
| BI460344.1 | - | 4 | 445 | |||
| U41745.1 | - | 4 | 340 | |||

PDAP1
Genomic Map