siRNA Details
| Gene Symbol: CYP2U1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183075.2 | NP_898898.1 | 3, 4 | 1370 | ||
| XM_005262717.2 | XP_005262774.1 | 3, 4 | 1436 | |||
| XM_005262720.2 | XP_005262777.1 | 2, 3 | 746 | |||
| GenBank mRNA | AK290002.1 | - | 3, 4 | 1375 | ||
| AK298989.1 | - | 2, 3 | 292 | |||
| AK303284.1 | - | 4, 5 | 1175 | |||
| AK304724.1 | - | 4, 5 | 1248 | |||
| AY343323.1 | - | 3, 4 | 1326 | |||
| BC132767.1 | - | 3, 4 | 1335 | 40146789 | ||
| BC136483.1 | - | 3, 4 | 1335 | 9020472 | ||

CYP2U1
Genomic Map