siRNA Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138431.2 | NP_612440.1 | 4 | 1290 | ||
| NR_130120.1 | 3 | 1142 | ||||
| XM_011516806.2 | XP_011515108.1 | 4 | 2237 | |||
| XM_017013005.1 | XP_016868494.1 | 4 | 2245 | |||
| GenBank mRNA | BC011982.1 | - | 4 | 1157 | 3860434 | |
| BF688676.1 | - | 3 | 416 | |||
| DB520468.1 | - | 2 | 144 | |||

MFSD3
Genomic Map