siRNA Details
| Gene Symbol: CHRNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000745.3 | NP_000736.2 | 5 | 1153 | ||
| GenBank mRNA | AF385586.1 | - | 5 | 1099 | ||
| BC033639.1 | - | 5 | 1115 | 5501542 | ||
| BC050469.1 | - | 1 | 929 | |||
| M83712.1 | - | 5 | 1101 | |||
| U62434.1 | - | 5 | 1107 | |||
| Y08419.1 | - | 5 | 953 | |||

CHRNA5
Genomic Map