siRNA Details
| Gene Symbol: TXNRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173513.1 | NP_001166984.1 | 6 | 824 | ||
| NM_052883.1 | NP_443115.1 | 6 | 824 | |||
| GenBank mRNA | AF133519.1 | - | 6 | 492 | ||
| AF171055.1 | - | 6 | 498 | |||
| AK299502.1 | - | 6 | 801 | |||
| BC030028.2 | - | 6 | 802 | |||
| BC050032.1 | - | 6 | 811 | |||

TXNRD3
Genomic Map