siRNA Details
| Gene Symbol: PKD1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076780.1 | NP_001070248.1 | 7 | 1406 | ||
| NM_052892.3 | NP_443124.3 | 7 | 1406 | |||
| NR_126532.1 | 7 | 1406 | ||||
| GenBank mRNA | AY164483.1 | - | 7 | 1406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKD1L2| Gene Symbol: PKD1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076780.1 | NP_001070248.1 | 7 | 1406 | ||
| NM_052892.3 | NP_443124.3 | 7 | 1406 | |||
| NR_126532.1 | 7 | 1406 | ||||
| GenBank mRNA | AY164483.1 | - | 7 | 1406 | ||