siRNA Details
| Gene Symbol: SLC25A25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006641.3 | NP_001006642.1 | 3 | 550 | ||
| NM_001006642.3 | NP_001006643.1 | 3 | 795 | |||
| NM_001265614.2 | NP_001252543.1 | 3 | 795 | |||
| NM_052901.4 | NP_443133.2 | 3 | 413 | |||
| NR_049766.2 | 3 | 456 | ||||
| XM_005251688.1 | XP_005251745.1 | 3 | 550 | |||
| XM_005251689.4 | XP_005251746.1 | 3 | 809 | |||
| XM_006716948.2 | XP_006717011.1 | 3 | 951 | |||
| XM_006716949.3 | XP_006717012.1 | 3 | 811 | |||
| GenBank mRNA | AB067483.1 | - | 3 | 794 | ||
| AJ619963.1 | - | 3 | 714 | |||
| AJ619989.1 | - | 3 | 338 | |||
| AJ619990.1 | - | 3 | 433 | |||
| AJ619991.1 | - | 3 | 395 | |||
| AJ619992.1 | - | 3 | 381 | |||
| AK290705.1 | - | 3 | 386 | |||
| AK290991.1 | - | 3 | 550 | |||
| AY358515.1 | - | 3 | 388 | |||
| BC089448.1 | - | 3 | 404 | 30558551 | ||
| BC103930.1 | - | 3 | 355 | 40008034 | ||
| BC103931.1 | - | 3 | 355 | 40008035 | ||
| BC103932.1 | - | 3 | 355 | 40008036 | ||
| BC103933.1 | - | 3 | 355 | 40008037 | ||
| BQ637100.1 | - | 3 | 413 | |||

SLC25A25
Genomic Map