siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052907.3 | NP_443139.2 | 5 | 1331 | ||
| XM_011537852.2 | XP_011536154.1 | 5 | 1388 | |||
| XM_017018767.1 | XP_016874256.1 | 6 | 1497 | |||
| GenBank mRNA | AB058689.1 | - | 4 | 1018 | ||

TMEM132B
Genomic Map