siRNA Details
| Gene Symbol: OSBPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144063.1 | NP_001137535.1 | 9 | 1045 | ||
| NM_020896.3 | NP_065947.1 | 10 | 1249 | |||
| NM_145638.2 | NP_663613.1 | 9 | 1038 | |||
| XM_011519873.2 | XP_011518175.1 | 10 | 1588 | |||
| XM_017017163.1 | XP_016872652.1 | 10 | 1596 | |||
| XM_017017164.1 | XP_016872653.1 | 5 | 1167 | |||
| GenBank mRNA | AB074006.1 | - | 10 | 1190 | ||
| AF392453.1 | - | 10 | 1162 | |||
| AF410430.1 | - | 9 | 1107 | |||
| AK055617.1 | - | 5 | 583 | |||
| AK056510.1 | - | 10 | 1207 | |||
| AK293000.1 | - | 9 | 994 | |||
| AK301003.1 | - | 8 | 885 | |||
| AK303205.1 | - | 10 | 1067 | |||
| AK309411.1 | - | 10 | 1227 | |||
| AL136918.1 | - | 10 | 1162 | |||
| BC032646.1 | - | 9 | 959 | 5518491 | ||
| BC039579.1 | - | 10 | 1160 | 5769002 | ||

OSBPL5
Genomic Map