siRNA Details
| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022776.4 | NP_073613.2 | 5, 6 | 1407 | ||
| GenBank mRNA | AF346292.1 | - | 5, 6 | 834 | ||
| AF392454.1 | - | 5, 6 | 978 | |||
| AK292702.1 | - | 5, 6 | 759 | |||
| BC065213.1 | - | 5, 6 | 757 | 3916115 | ||

OSBPL11
Genomic Map