siRNA Details
| Gene Symbol: TMEM123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052932.2 | NP_443164.2 | 3 | 695 | ||
| GenBank mRNA | AK075420.1 | - | 3 | 479 | ||
| AK225790.1 | - | 3 | 666 | |||
| AY008283.1 | - | 2 | 433 | |||
| AY157580.1 | - | 3 | 390 | |||
| AY358696.1 | - | 3 | 367 | |||
| BC032296.1 | - | 3 | 382 | 4340763 | ||

TMEM123
Genomic Map