siRNA Details
| Gene Symbol: PTPMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143984.1 | NP_001137456.1 | 2 | 633 | ||
| NM_175732.2 | NP_783859.1 | 4 | 716 | |||
| GenBank mRNA | AF277187.1 | - | 2 | 130 | ||
| AK294640.1 | - | 3 | 355 | |||
| AY333987.1 | - | 3 | 331 | |||
| BC014048.1 | - | 3 | 362 | |||
| BC020242.1 | - | 4 | 545 | 4591438 | ||
| BC107573.1 | - | 2 | 468 | |||
| BQ050653.1 | - | 2 | 109 | |||
| DR004030.1 | - | 4 | 574 | |||

PTPMT1
Genomic Map