siRNA Details
| Gene Symbol: FBXO17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024907.6 | NP_079183.4 | 3 | 693 | ||
| NM_148169.2 | NP_680474.1 | 3 | 580 | |||
| GenBank mRNA | AF386743.1 | - | 3 | 620 | ||
| AK057934.1 | - | 3 | 580 | |||
| BC012385.1 | - | 3 | 531 | 3864760 | ||

FBXO17
Genomic Map