siRNA Details
| Gene Symbol: FBXO17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024907.6 | NP_079183.4 | 4 | 719 | ||
| NM_148169.2 | NP_680474.1 | 4 | 606 | |||
| NR_104026.1 | 4 | 757 | ||||
| GenBank mRNA | AF386743.1 | - | 4 | 646 | ||
| AK021860.1 | - | 1 | 160 | |||
| AK057934.1 | - | 4 | 606 | |||
| BC012385.1 | - | 4 | 557 | 3864760 | ||
| BX338290.2 | - | 4 | 646 | |||

FBXO17
Genomic Map