siRNA Details
| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 704 | ||
| GenBank mRNA | BC013062.1 | - | 1 | 655 | 2901027 | |
| BI757526.1 | - | 1 | 661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM122A| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 704 | ||
| GenBank mRNA | BC013062.1 | - | 1 | 655 | 2901027 | |
| BI757526.1 | - | 1 | 661 | |||