siRNA Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160407.1 | NP_001153879.1 | 4 | 557 | ||
| NM_138463.3 | NP_612472.1 | 4 | 690 | |||
| XM_006721671.3 | XP_006721734.1 | 4 | 493 | |||
| GenBank mRNA | BC014072.2 | - | 4 | 686 | 3957355 | |
| CA453962.1 | - | 4 | 696 | |||

TLCD1
Genomic Map