siRNA Details
| Gene Symbol: RBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052960.2 | NP_443192.1 | 1, 2 | 141 | ||
| GenBank mRNA | AF399927.1 | - | 1, 2 | 122 | ||
| AK312027.1 | - | 1, 2 | 109 | |||
| AY145438.1 | - | 1, 2 | 108 | |||
| BC063013.1 | - | 1, 2 | 141 | 5224517 | ||

RBP7
Genomic Map