siRNA Details
| Gene Symbol: RBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052960.2 | NP_443192.1 | 3, 4 | 410 | ||
| GenBank mRNA | AF399927.1 | - | 3, 4 | 391 | ||
| AK312027.1 | - | 3, 4 | 378 | |||
| AY145438.1 | - | 3, 4 | 377 | |||
| BC033883.1 | - | 4, 5 | 623 | |||
| BC063013.1 | - | 3, 4 | 410 | 5224517 | ||

RBP7
Genomic Map