siRNA Details
| Gene Symbol: TM4SF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184723.1 | NP_001171652.1 | 1 | 110 | ||
| NM_138786.3 | NP_620141.1 | 2 | 286 | |||
| GenBank mRNA | AK313400.1 | - | 2 | 60 | ||
| AL833134.1 | - | 2 | 209 | |||
| BC014339.1 | - | 2 | 162 | 3828466 | ||
| BF213446.1 | - | 121 | ||||
| CA454331.1 | - | 1 | 72 | |||
| DB282139.1 | - | 2 | 286 | |||

TM4SF18
Genomic Map