siRNA Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053050.4 | NP_444278.1 | 3 | 398 | ||
| GenBank mRNA | BC012163.1 | - | 3 | 249 | 4622995 | |
| BG196300.1 | - | 3 | 383 | |||
| BQ014848.1 | - | 1 | 244 | |||
| BQ688110.1 | - | 3 | 255 | |||

MRPL53
Genomic Map