siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 9 | 1072 | ||
| XM_017010236.1 | XP_016865725.1 | 9 | 1263 | |||
| GenBank mRNA | AJ420537.1 | - | 3 | 198 | ||
| AK124442.1 | - | 9 | 951 | |||
| AK289918.1 | - | 9 | 1091 | |||
| AK293853.1 | - | 8 | 878 | |||
| BC069567.1 | - | 5 | 475 | |||
| BC069570.1 | - | 9 | 992 | 7262298 | ||

SLC18B1
Genomic Map