siRNA Details
| Gene Symbol: CATSPER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282309.1 | NP_001269238.1 | 3 | 173 | ||
| NM_001282310.1 | NP_001269239.1 | 2 | 88 | |||
| NM_054020.3 | NP_473361.1 | 2 | 274 | |||
| NM_172095.2 | NP_742093.1 | 2 | 274 | |||
| NR_110319.1 | 1 | 61 | ||||
| XM_017021904.1 | XP_016877393.1 | 2 | 280 | |||
| GenBank mRNA | AF411817.1 | - | 1 | 59 | ||
| AF411818.1 | - | 1 | 59 | |||
| AF411819.1 | - | 1 | 59 | |||
| AK122607.1 | - | 3 | 172 | |||
| BC028728.1 | - | 2 | 279 | |||
| BC064387.1 | - | 2 | 88 | 5744415 | ||
| BX648099.1 | - | 2 | 310 | |||
| Gene Symbol: CATSPER2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002318.2 | 2 | 315 | |||
| GenBank mRNA | BC047442.1 | - | 2 | 314 | ||
| BC066967.1 | - | 2 | 315 | |||

CATSPER2
Genomic Map