siRNA Details
| Gene Symbol: SH2D1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053282.4 | NP_444512.2 | 1 | 170 | ||
| GenBank mRNA | AF256653.1 | - | 1 | 48 | ||
| AF403479.1 | - | 1 | 48 | |||
| AF484964.1 | - | 1 | 146 | |||
| AK097162.1 | - | 1 | 97 | |||
| AK314743.1 | - | 1 | 117 | |||
| AL709120.1 | - | 1 | 170 | |||
| BC022407.1 | - | 1 | 158 | 4717041 | ||
| BC066595.1 | - | 1 | 144 | 5755414 | ||

SH2D1B
Genomic Map