siRNA Details
Gene Symbol: GPR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080865.3 | NP_543141.3 | 1 | 1094 | ||
GenBank mRNA | AB032600.1 | - | 1 | 1060 | ||
BC026357.1 | - | 1 | 1027 | 4812283 | ||
HQ709188.1 | - | 1 | 755 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
